missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl-1,2-epoxybutane-d5, TRC

Product Code. 30355338
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30355338 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30355338 Supplier Toronto Research Chemicals Supplier No. TRCC37952225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Chlorophenyl-1,2-epoxybutane-d5
CAS 1189717-28-9
Molekylær formel C10 2H5 H6 Cl O
Mængde 25 mg
Synonym 2-[2-(4-Chlorophenyl)-1,1-dideuterioethyl]-2,3,3-trideuteriooxirane, 4-Chlorophenyl-1,2-epoxybutane-d5, 4-(4-Chlorophenyl)-1,2-epoxybutane-1,1,2,3,3-d5
InChI formel InChI=1S/C10H11ClO/c1-2-9-10(12-9)7-3-5-8(11)6-4-7/h3-6,9-10H,2H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C1OC1c2ccc(Cl)cc2
IUPAC navn 2-(4-chlorophenyl)-3-(1,1,2,2,2-pentadeuterioethyl)oxirane
Molekylvægt (g/mol) 187.68
Formel vægt 187.0812
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.