Få mere at vide
(4-Chlorophenoxy)acetic Acid Ethyl Ester, Mikromol™
Beskrivelse
(4-Chlorophenoxy)acetic Acid Ethyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (4-Chlorophenoxy)acetic Acid Ethyl Ester |
| CAS | 14426-42-7 |
| Analyt- eller komponentnavne | (4-Chlorophenoxy)acetic Acid Ethyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H11 Cl O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-(4-Chlorophenoxy)acetic Acid Ethyl Ester, Acetic acid, (4-chlorophenoxy)-, ethyl ester (9CI), Acetic acid, (p-chlorophenoxy)-, ethyl ester (6CI,7CI,8CI), (4-Chlorophenoxy)acetic acid ethyl ester, Ethyl (4-chlorophenoxy)acetate, Ethyl (p-chlorophenoxy)acetate, Ethyl 2-(4-chlorophenoxy)acetate, Ethyl 2-(4-chlorophenoxy)acetate, Kangailing, NSC 66307, (4-Chlorophenoxy)acetic Acid Ethyl Ester |
| InChI formel | InChI=1S/C10H11ClO3/c1-2-13-10(12)7-14-9-5-3-8(11)4-6-9/h3-6H,2,7H2,1H3 |
| SMIL | CCOC(=O)COc1ccc(Cl)cc1 |
| IUPAC navn | ethyl 2-(4-chlorophenoxy)acetate |
| Vis mere |