Learn More
4-Chlorophenol, Sigma-Aldrich™
Product Code. 15838720
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15838720 1 kg 1kg
15818720 100 g 100g
15828720 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15838720 Supplier Sigma Aldrich Supplier No. 8022541000
 Restricted Item
Forventet forsendelse: 24-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-48-9
Grade Research
Molecular Formula 4-(Cl)C6H4OH
Molecular Weight (g/mol) 128.56
Specifications
Density 1.26 g/cm3 at 45°C
Boiling Point 220°C/1013 hPa
MDL Number MFCD00002318
Quantity 1 kg
Synonym 4-Chlorophenol, 4-Chloro-1-hydroxybenzene, 4-Chlorophenol