Få mere at vide
4-Chloronitrobenzene-d4, TRC
Artikelnummer. 30351416
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30351416 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351416 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3739121G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloronitrobenzene-d4
CAS 68239-23-6
Molekylær formel C6 D4 Cl N O2
Mængde 1 g
Synonym 1-Chloro-4-nitrobenzene-d4, 1-Nitro-4-chlorobenzene-d4, 4-Chloro-1-nitrobenzene-d4, 4-Nitro-1-chlorobenzene-d4, 4-Nitrochlorobenzene-d4, 4-Nitrophenyl-d4 Chloride, NSC 9792-d4, PNCB-d4, p-Chloronitrobenzene-d4, p-Nitrochlorobenzene-d4, p-Nitrophenyl-d4 Chloride
InChI formel InChI=1S/C6H4ClNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1Cl)[N+](=O)[O-]
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-nitrobenzene
Molekylvægt (g/mol) 161.58
Formel vægt 161.0182