Få mere at vide
4-Chloronitrobenzene-d4, CDN
Artikelnummer. 30324729
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324729 0.5 g
30317845 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324729 Leverandør CDN Leverandørnr. CDND54690.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloronitrobenzene-d4
CAS 68239-23-6
Molekylær formel C6 D4 Cl N O2
Mængde 0.5 g
Synonym 1-Chloro-4-nitrobenzene-d4, 1-Nitro-4-chlorobenzene-d4, 4-Chloro-1-nitrobenzene-d4, 4-Nitro-1-chlorobenzene-d4, 4-Nitrochlorobenzene-d4, 4-Nitrophenyl-d4 Chloride, NSC 9792-d4, PNCB-d4, p-Chloronitrobenzene-d4, p-Nitrochlorobenzene-d4, p-Nitrophenyl-d4 Chloride
InChI formel InChI=1 S/C6H4ClNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cl)[N+](=O)[O-]
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-nitrobenzene
Molekylvægt (g/mol) 161.58
Formel vægt 161.0182 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity