missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroisatin, TRC

Product Code. 30365189
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30365189 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365189 Supplier Toronto Research Chemicals Supplier No. TRCC428602100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Chloroisatin
CAS 6344-05-4
Molekylær formel C8H4ClNO2
Mængde 100 mg
Synonym 4-Chloro-1H-indole-2,3-dione, 4-Chloroindoline-2,3-dione, 4-Chloroisatin, NSC 49794
InChI formel InChI=1S/C8H4ClNO2/c9-4-2-1-3-5-6(4)7(11)8(12)10-5/h1-3H,(H,10,11,12)
SMIL Clc1cccc2NC(=O)C(=O)c12
IUPAC navn 4-chloro-1H-indole-2,3-dione
Molekylvægt (g/mol) 181.58
Formel vægt 180.9931
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.