missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyronitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-7024383
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
628-20-6
C4H6ClN
103.55
MFCD00001972
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
12336
4-chlorobutanenitrile
ClCCCC#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
628-20-6
C4H6ClN
103.55
MFCD00001972
ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
12336
4-chlorobutanenitrile
ClCCCC#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.084
Boiling Point 195°C to 197°C
Flash Point 85°C (185°F)
Linear Formula Cl(CH2)3CN
Refractive Index 1.444
Quantity 50 g
UN Number UN3276
Beilstein 1633582
Formula Weight 103.55
Percent Purity 97%
Chemical Name or Material 4-Chlorobutyronitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'