Få mere at vide
4-Chlorobutyric anhydride, TRC
Artikelnummer. 30356409
Skift visning
Mængde:
250 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30356409 1 g -
30360880 250 mg -
30366122 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30356409 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2929551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20496-01-9
Molekylær formel C8H12Cl2O3
Molekylvægt (g/mol) 227.09
Synonym Butanoic acid, 4-chloro-, anhydride (9CI), Butyric acid, 4-chloro-, anhydride (7CI,8CI), 4-Chlorobutanoic anhydride
IUPAC navn 4-chlorobutanoyl 4-chlorobutanoate
SMIL ClCCCC(=O)OC(=O)CCCCl
Tekniske data
Kemisk navn eller materiale 4-Chlorobutyric anhydride
Mængde 1 g
InChI formel InChI=1S/C8H12Cl2O3/c9-5-1-3-7(11)13-8(12)4-2-6-10/h1-6H2
Formel vægt 226.0163