Få mere at vide
4-Chlorobiphenyl-2',3',4',5',6'-d5, CDN
Artikelnummer. 30323235
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323235 0.25 g -
30314329 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323235 Leverandør CDN Leverandørnr. CDND68980.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorobiphenyl-2',3',4',5',6'-D5
CAS 51624-37-4
Molekylær formel C12 D5 H4 Cl
Mængde 0.25 g
Synonym 1,1'-Biphenyl-2,3,4,5,6-d5, 4'-chloro- (9 CI), 4'-Chloro-1,1'-biphenyl-2,3,4,5,6-d5, 4-Chlorobiphenyl-2',3',4',5',6'-d5, 4-Chlorobiphenyl-2',3',4',5',6'-D5, PCB No. 3 D5, PCB 3-d5
InChI formel InChI=1 S/C12H9Cl/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9 H/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2ccc(Cl)cc2
IUPAC navn 1-(4-chlorophenyl)-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 193.6837
Formel vægt 193.0707 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity