missing translation for 'onlineSavingsMsg'
Learn More

4-chlorobenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number 10454795
Click to view available options
Quantity:
2.5 g
25 g
50 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104-86-9
C7H8ClN
141.6
MFCD00008121
YMVFJGSXZNNUDW-UHFFFAOYSA-N
4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
66036
(4-chlorophenyl)methanamine
C1=CC(=CC=C1CN)Cl
This item is not returnable. View return policy
104-86-9
C7H8ClN
141.6
MFCD00008121
YMVFJGSXZNNUDW-UHFFFAOYSA-N
4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
66036
(4-chlorophenyl)methanamine
C1=CC(=CC=C1CN)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1600g/mL
Boiling Point 215.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2NH2
Refractive Index 1.5576 to 1.5610
Quantity 25 g
Beilstein 12, 1074
Specific Gravity 1.16
Solubility Information Solubility in water: immiscible with water
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-chlorobenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chlorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'