Learn More
4-Chlorobenzylamine, 97+%, Thermo Scientific Chemicals
Catalog Number p-7021585
Click to view available options
Quantity:
5 g
25 g
100 g
104-86-9
C7H8ClN
141.598
MFCD00008121
YMVFJGSXZNNUDW-UHFFFAOYSA-N
4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
66036
(4-chlorophenyl)methanamine
C1=CC(=CC=C1CN)Cl
104-86-9
C7H8ClN
141.598
MFCD00008121
YMVFJGSXZNNUDW-UHFFFAOYSA-N
4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
66036
(4-chlorophenyl)methanamine
C1=CC(=CC=C1CN)Cl
4-Chlorobenzylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorobenzylamine is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY
Density 1.16
Boiling Point 229°C
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.559
Quantity 5 g
UN Number UN2735
Beilstein 507949
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 141.6
Percent Purity ≥97%
Chemical Name or Material 4-Chlorobenzylamine