Få mere at vide
4-Chlorobenzonitrile-d4, CDN
Artikelnummer. 30318446
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318446 0.1 g 0.1g
30322405 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318446 Leverandør CDN Leverandørnr. CDND65760.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorobenzonitrile-D4
CAS 1219794-82-7
Molekylær formel C7 D4 Cl N
Mængde 0.1 g
Synonym Benzonitrile-2,3,5,6-d4, 4-chloro- (ACI), 4-Chlorobenzonitrile-2,3,5,6-d4 (ACI), 4-Chlorobenzonitrile-d4, 4-Chlorobenzonitrile-D4, 1-Chloro-4-cyanobenzene-2,3,5,6-d4
InChI formel InChI=1 S/C7H4ClN/c8-7-3-1-6(5-9)2-4-7/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C#N)c([2 H])c([2 H])c1Cl
IUPAC navn 4-chloro-2,3,5,6-tetradeuteriobenzonitrile
Molekylvægt (g/mol) 141.591
Formel vægt 141.0283 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity