Få mere at vide
4-Chlorobenzoic-d4 Acid, CDN
Artikelnummer. 30322292
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322292 0.5 g 0.5g
30319466 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322292 Leverandør CDN Leverandørnr. CDND50250.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorobenzoic acid ring-d4
CAS 85577-25-9
Molekylær formel C7 2H4 H Cl O2
Mængde 0.5 g
InChI formel InChI=1 S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c([2 H])c1Cl
IUPAC navn 4-chloro-2,3,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 160.59
Formel vægt 160.0229 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity