Få mere at vide
4-Chlorobenzoic Acid-d4, TRC
Artikelnummer. 30367014
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30367014 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367014 Leverandør Toronto Research Chemicals Leverandørnr. TRCC364652100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorobenzoic acid ring-d4
CAS 85577-25-9
Molekylær formel C7 2H4 H Cl O2
Mængde 100 mg
Synonym Benzoic-2,3,5,6-d4 acid, 4-chloro- (9CI), 4-Chlorobenzoic Acid-d4, 4-Chlorobenzoic Acid-D4, 4-Chlorobenzoic-d4 Acid, 4-Chlorobenzoic-2,3,5,6-d4 Acid, p-Chlorobenzoic Acid-d4
InChI formel InChI=1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1Cl
IUPAC navn 4-chloro-2,3,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 160.59
Formel vægt 160.0229