missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7052816
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
195062-61-4
C12H16BClO2
238.52
MFCD05663875
NYARTXMDWRAVIX-UHFFFAOYSA-N
10633712
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
195062-61-4
C12H16BClO2
238.52
MFCD05663875
NYARTXMDWRAVIX-UHFFFAOYSA-N
10633712
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 50°C to 55°C
Color Brown to White
Quantity 1 g
Flash Point >110°C (230°F)
Physical Form Solid
Synonym 2-4-chlorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chlorophenylboronic acid pinacol ester, 4-chlorophenylboronic acid, pinacol ester, 1,3,2-dioxaborolane,2-4-chlorophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chlorobenzene, 4-chlorophenyl boronic acid pinacol ester, 4-chlorobenzeneboronic acid, pinacol ester, p-chlorophenylboronic acid 2,3-dimethyl-2,3-butanediyl
Formula Weight 238.52
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'