Få mere at vide
4-Chlorobenzene-d4-sulfonamide, TRC
Artikelnummer. 30365582
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30365582 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365582 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36451225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorobenzene-d4-sulfonamide
CAS 544686-14-8
Molekylær formel C6 2H4 H2 Cl N O2 S
Mængde 25 mg
InChI formel InChI=1S/C6H6ClNO2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,(H2,8,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1Cl)S(=O)(=O)N
IUPAC navn 4-chloro-2,3,5,6-tetradeuteriobenzenesulfonamide
Molekylvægt (g/mol) 195.66
Formel vægt 195.0059