Få mere at vide
4-Chloroanisole-2,3,5,6-d4, CDN
Artikelnummer. 30310395
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310395 0.5 g 0.5g
30317516 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310395 Leverandør CDN Leverandørnr. CDND61050.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloroanisole-2,3,5,6-D4
CAS 1219804-86-0
Molekylær formel C7 D4 H3 Cl O
Mængde 0.5 g
Synonym 3-Chloro-6-methoxybenzene-1,2,4,5-d4, 1-Chloro-4-methoxy-2,3,5,6-tetradeuteriobenzene, 4-Chloroanisole-d4, 4-Chloroanisole-2,3,5,6-d4, 4-Chloroanisole-2,3,5,6-D4, 1-Chloro-4-methoxybenzene-2,3,5,6-d4
InChI formel InChI=1 S/C7H7ClO/c1-9-7-4-2-6(8)3-5-7/h2-5 H,1H3/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(OC)c([2 H])c([2 H])c1Cl
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-methoxybenzene
Molekylvægt (g/mol) 146.6075
Formel vægt 146.0436 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity