missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Chloroaniline, Sigma-Aldrich™

Product Code. 15898790
Change view
Click to view available options
Mængde:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
15898790 100 g
15808800 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15898790 Supplier Sigma Aldrich Supplier No. 8026130100
 Begrænset vare
Forventet forsendelse: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-47-8
Grad Research
Molekylvægt (g/mol) 127.57

Specifications

Smeltepunkt 66°C to 69°C
Tæthed 1.17 g/cm3 at 70°C
Kogepunkt 232°C/1013 hPa
MDL nummer MFCD00007835
Mængde 100 g
Synonym 4-Chloroaniline, p-Chloroaniline, 4-Chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.