Få mere at vide
4-Chloroaniline-2,3,5,6-d4, CDN
Artikelnummer. 30320708
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320708 0.1 g 0.1g
30319750 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320708 Leverandør CDN Leverandørnr. CDND24610.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloroaniline-d4
CAS 191656-33-4
Molekylær formel C6 D4 H2 Cl N
Mængde 0.1 g
Synonym Benzen-2,3,5,6-d4-amine, 4-chloro- (9 CI), 4-Chlorobenzen-2,3,5,6-d4-amine, 4-Chloroaniline-d4, 4-Chloroaniline-2,3,5,6-D4
InChI formel InChI=1 S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4 H,8H2/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(Cl)c([2 H])c([2 H])c1N
IUPAC navn 4-chloro-2,3,5,6-tetradeuterioaniline
Molekylvægt (g/mol) 131.5962
Formel vægt 131.044 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity