Få mere at vide
4-Chloro Trazodone Isomer, TRC
Artikelnummer. 30180213
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180213 25 mg
30179889 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30180213 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42165025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 157072-19-0
Molekylær formel C19 H22 Cl N5 O
Molekylvægt (g/mol) 371.8639
Synonym 1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 2-[3-[4-(4-chlorophenyl)-1-piperazinyl]propyl]-, 2-[3-[4-(4-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one, 4-Chlorotrazodone, 4-Chloro trazodone, 4-Chloro Trazodone Isomer
IUPAC navn 2-[3-[4-(4-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one
SMIL Clc1ccc(cc1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2
Tekniske data
Kemisk navn eller materiale 2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one
Mængde 25 mg
InChI formel InChI=1S/C19H22ClN5O/c20-16-5-7-17(8-6-16)23-14-12-22(13-15-23)9-3-11-25-19(26)24-10-2-1-4-18(24)21-25/h1-2,4-8,10H,3,9,11-15H2
Formel vægt 371.1513