missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-phenylbenzamide, 97%

Brand:  Alfa Aesar™ H59590.03

Additional Details : CAS Number : 6833-15-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485958

  • 1707.87 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

6833-15-4
231.679
SFHDVPIEJXCMBP-UHFFFAOYSA-N
346669
C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl
C13H10ClNO
MFCD00000620
4-chlorobenzanilide, benzamide,4-chloro-n-phenyl, benzamide, 4-chloro-n-phenyl, p-chlorobenzanilide, 4-chlor-benzanilid, benzanilide, 4-chloro, n-phenyl-4-chlorobenzamide, 4-chloro-n-phenyl-benzamide, 4-chlorophenyl-n-benzamide, 4-chloranyl-n-phenyl-benzamide
4-chloro-N-phenylbenzamide

Specifications

4-Chloro-N-phenylbenzamide
6833-15-4
MFCD00000620
SFHDVPIEJXCMBP-UHFFFAOYSA-N
4-chloro-N-phenylbenzamide
346669
Solid
1g
C13H10ClNO
4-chlorobenzanilide, benzamide,4-chloro-n-phenyl, benzamide, 4-chloro-n-phenyl, p-chlorobenzanilide, 4-chlor-benzanilid, benzanilide, 4-chloro, n-phenyl-4-chlorobenzamide, 4-chloro-n-phenyl-benzamide, 4-chlorophenyl-n-benzamide, 4-chloranyl-n-phenyl-benzamide
C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl
231.679
231.68
97%
Videos
SDS Documents