Learn More
4-Chloro-N,N-diisopropylbenzamide, 97%, Thermo Scientific™
Catalog Number 15436268
Click to view available options
Quantity:
250 mg
1 g
79606-45-4
C13H18ClNO
239.743
MFCD00032340
MGPPJDHMGGQMSG-UHFFFAOYSA-N
4-chloro-n,n-diisopropylbenzamide, 4-chloro-n,n-di propan-2-yl benzamide, acmc-1be6x, n,n-diisopropyl-4-chlorobenzamide, 4-chloro-n,n-bis propan-2-yl benzamide
241026
4-chloro-N,N-di(propan-2-yl)benzamide
CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)Cl
79606-45-4
C13H18ClNO
239.743
MFCD00032340
MGPPJDHMGGQMSG-UHFFFAOYSA-N
4-chloro-n,n-diisopropylbenzamide, 4-chloro-n,n-di propan-2-yl benzamide, acmc-1be6x, n,n-diisopropyl-4-chlorobenzamide, 4-chloro-n,n-bis propan-2-yl benzamide
241026
4-chloro-N,N-di(propan-2-yl)benzamide
CC(C)N(C(C)C)C(=O)C1=CC=C(C=C1)Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 239.75
Percent Purity 97%
Chemical Name or Material 4-Chloro-N,N-diisopropylbenzamide