Få mere at vide
4-Chloro-N-(methyl-d3)pyridine-2-carboxamide, TRC
Artikelnummer. 30349438
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30349438 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349438 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37995250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloro-N-(methyl-d3)pyridine-2-carboxamide
CAS 1189858-48-7
Molekylær formel C7H4D3ClN2O
Mængde 50 mg
Synonym 4-Chloro-N-(methyld3)-2-pyridinecarboxamide, 4-Chloro-N-(methyl-d3)picolinamide, 4-Chloropyridine-2-carboxylic Acid (Methyl-d3)amide, N-(Methyl-d3)-4-chloro-2-pyridinecarboxamide
InChI formel InChI=1S/C7H7ClN2O/c1-9-7(11)6-4-5(8)2-3-10-6/h2-4H,1H3,(H,9,11)/i1D3
SMIL ClC1=CC=NC(C(NC([2H])([2H])[2H])=O)=C1
IUPAC navn 4-chloro-N-(methyl-d3)picolinamide
Molekylvægt (g/mol) 173.61
Formel vægt 173.04