missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-3-(trifluoromethyl)benzamide, 95%

Brand:  Alfa Aesar™ H65412.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469626

  • 1908.81 DKK / 1g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27975760

Specifications

4-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-3-(trifluoromethyl)benzamide
MFCD27975760
Solid
1g
353.77
95%
Videos
SDS