missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-methoxyphenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65179.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469436

  • 2009.39 DKK / 1g


Estimated Shipment date
from Supplier 17-10-2018
Add to basket

Specifications

Chemical Identifiers

MFCD28098764

Specifications

4-Chloro-N-(4-methoxyphenyl)-3-sulfamoylbenzamide
95%
340.78
1g
MFCD28098764
Solid
Videos
SDS