Learn More
Alfa Aesar™ 4-Chloro-N-(4-chloro-2-methylphenyl)-3-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65809.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469916
  • 1908.81 DKK / 1g
Estimated Shipment date
from Supplier 15-01-2019
Add to basket
Specifications
Chemical Identifiers
MFCD28098828
Specifications
4-Chloro-N-(4-chloro-2-methylphenyl)-3-sulfamoylbenzamide
MFCD28098828
Solid
1g
359.23
95%