missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-(4-chloro-2-fluorophenyl)-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65560.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469736

  • 1908.81 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27975785

Specifications

4-Chloro-N-(4-chloro-2-fluorophenyl)-3-sulfamoylbenzamide
MFCD27975785
Solid
1g
363.19
95%
Videos
SDS