Få mere at vide
4-Chloro-N-(2-hydroxyethyl)benzamide, Mikromol™
Artikelnummer. 30051128
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051128 100 mg
30049483 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051128 Leverandør Mikromol Leverandørnr. MM0250.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Chloro-N-(2-hydroxyethyl)benzamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloro-N-(2-hydroxyethyl)benzamide
CAS 7400-54-6
Analyt- eller komponentnavne 4-Chloro-N-(2-hydroxyethyl)benzamide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H10 Cl N O2
Certificeringer/compliance ISO 17025
Synonym 4-Chloro-N-(2-hydroxyethyl)-benzamide, Benzamide, p-chloro-N-(2-hydroxyethyl)- (6CI,7CI), 2-(4-Chlorobenzoylamino)ethanol, 4-Chloro-N-(2-hydroxyethyl)benzamide, NSC 33145, Ro 18-1675
InChI formel InChI=1S/C9H10ClNO2/c10-8-3-1-7(2-4-8)9(13)11-5-6-12/h1-4,12H,5-6H2,(H,11,13)
SMIL OCCNC(=O)c1ccc(Cl)cc1
IUPAC navn 4-chloro-N-(2-hydroxyethyl)benzamide
Molekylvægt (g/mol) 199.63
Mængde 100 mg
Formel vægt 199.04 g/mol