Få mere at vide
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide-d4, TRC
Artikelnummer. 30354012
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30354012 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354012 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3666622MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide-d4
CAS 1189464-84-3
Molekylær formel C15H10D4ClNO2
Mængde 2 mg
InChI formel InChI=1S/C15H14ClNO2/c16-13-5-3-12(4-6-13)15(19)17-10-9-11-1-7-14(18)8-2-11/h1-8,18H,9-10H2,(H,17,19)/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(C(=O)NCCc2ccc(O)cc2)c([2H])c([2H])c1Cl
IUPAC navn 4-chloro-2,3,5,6-tetradeuterio-N-[2-(4-hydroxyphenyl)ethyl]benzamide
Molekylvægt (g/mol) 279.75
Formel vægt 279.0964