Learn More
Alfa Aesar™ 4-Chloro-N-(2,4-dimethylphenyl)-2-fluoro-5-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65487.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469676
  • 2009.39 DKK / 1g
Estimated Shipment date
from Supplier 08-06-2018
Add to basket
Specifications
Chemical Identifiers
4-Chloro-N-(2,4-dimethylphenyl)-2-fluoro-5-sulfamoylbenzamide
356.8
MFCD28098799
Specifications
4-Chloro-N-(2,4-dimethylphenyl)-2-fluoro-5-sulfamoylbenzamide
Solid
356.8
1g
95%
MFCD28098799