Få mere at vide
4-Chloro-L-phenylalanine Hydrochloride, TRC
Artikelnummer. 30178990
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30178990 500 mg
30178960 250 mg
30178983 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30178990 Leverandør Toronto Research Chemicals Leverandørnr. TRCC382308500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 123053-23-6
Molekylær formel C9H10ClNO2 . HCl
Molekylvægt (g/mol) 236.09
Synonym (L)-4-Chlorophenylalanine Hydrochloride;
IUPAC navn (2S)-2-amino-3-(4-chlorophenyl)propanoic acid;hydrochloride
SMIL Cl.N[C@@H](Cc1ccc(Cl)cc1)C(=O)O
Tekniske data
Kemisk navn eller materiale 4-Chloro-L-phenylalanine Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C9H10ClNO2.ClH/c10-7-3-1-6(2-4-7)5-8(11)9(12)13;/h1-4,8H,5,11H2,(H,12,13);1H/t8-;/m0./s1