missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-chloro-7-methylthieno[3,2-d]pyrimidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Synonym | 4-chloro-7-methylthieno 3,2-d pyrimidine, thieno 3,2-d pyrimidine, 4-chloro-7-methyl, 4-chloro-7-methylthiopheno 3,2-d pyrimidine, thieno 3,2-d pyrimidine,4-chloro-7-methyl, pubchem23125, 4-chloro-7-methyl thieno 3,2-d pyrimidine, 4-chloro-7-methyl-thieno 3,2-d pyrimidine, 4-chloranyl-7-methyl-thieno 3,2-d pyrimidine, 4-chloro-7-methylthieno 2,3-d pyrimidine |
Formula Weight | 184.65 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-7-methylthieno[3, 2-d]pyrimidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'