Få mere at vide
4-Chloro-7-methoxy-2-methylquinoline, TRC
Artikelnummer. 30366246
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366246 100 mg
30365826 500 mg
30360441 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366246 Leverandør Toronto Research Chemicals Leverandørnr. TRCC596955100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75896-68-3
Molekylær formel C11H10NOCl
Molekylvægt (g/mol) 207.66
IUPAC navn 4-chloro-7-methoxy-2-methylquinoline
SMIL COc1ccc2c(Cl)cc(C)nc2c1
Tekniske data
Kemisk navn eller materiale 4-Chloro-7-methoxy-2-methylquinoline
Mængde 100 mg
InChI formel InChI=1S/C11H10ClNO/c1-7-5-10(12)9-4-3-8(14-2)6-11(9)13-7/h3-6H,1-2H3
Formel vægt 207.0451