Få mere at vide
4-Chloro-6-(trifluoromethyl)-3-pyridinecarboxylic Acid, TRC
Artikelnummer. 30360920
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360920 500 mg
30364089 100 mg
30352595 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30360920 Leverandør Toronto Research Chemicals Leverandørnr. TRCC253066500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1060810-66-3
Molekylær formel C7H3NO2F3Cl
Molekylvægt (g/mol) 225.55
IUPAC navn 4-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMIL OC(=O)c1cnc(cc1Cl)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 4-Chloro-6-(trifluoromethyl)-3-pyridinecarboxylic Acid
Mængde 500 mg
InChI formel InChI=1S/C7H3ClF3NO2/c8-4-1-5(7(9,10)11)12-2-3(4)6(13)14/h1-2H,(H,13,14)
Formel vægt 224.9804