missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine, 96%, Thermo Scientific™

Catalog Number 15478365
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
312584-53-5
C13H9ClN2OS
276.738
MFCD01831011
MZBKUNFWNKYKSK-UHFFFAOYSA-N
4-chloro-5-4-methoxyphenyl thieno 2,3-d pyrimidine, 4-chloro-5-4-methoxy-phenyl-thieno 2,3-d-pyrimidine, 4-chloro-5-4-methoxyphenyl-thieno 2,3-d pyrimidine, 1-4-chlorothiopheno 2,3-d pyrimidin-5-yl-4-methoxybenzene, 4-chloro-5-4-methoxy-phenyl-thieno 2,3-d pyrimidine
708734
4-chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine
COC1=CC=C(C=C1)C2=CSC3=C2C(=NC=N3)Cl
This item is not returnable. View return policy
312584-53-5
C13H9ClN2OS
276.738
MFCD01831011
MZBKUNFWNKYKSK-UHFFFAOYSA-N
4-chloro-5-4-methoxyphenyl thieno 2,3-d pyrimidine, 4-chloro-5-4-methoxy-phenyl-thieno 2,3-d-pyrimidine, 4-chloro-5-4-methoxyphenyl-thieno 2,3-d pyrimidine, 1-4-chlorothiopheno 2,3-d pyrimidin-5-yl-4-methoxybenzene, 4-chloro-5-4-methoxy-phenyl-thieno 2,3-d pyrimidine
708734
4-chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine
COC1=CC=C(C=C1)C2=CSC3=C2C(=NC=N3)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 166°C to 168°C
Quantity 1 g
Formula Weight 276.75
Percent Purity 96%
Chemical Name or Material 4-Chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'