Learn More
4-Chloro-4'-hydroxybiphenyl, TRC
Product Code. 30360400
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360400 10 mg 10mg
30354393 100 mg 100mg
30362860 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30360400 Supplier Toronto Research Chemicals Supplier No. TRCC58583510MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28034-99-3
Molecular Formula C12H9OCl
Molecular Weight (g/mol) 204.65
Synonym 4-Hydroxy-4'-chlorobiphenyl, 4'-Chloro-4-biphenylol, 4'-Hydroxy-4-chlorobiphenyl, p-Chloro-p'-hydroxybiphenyl, 4'-Chloro[1,1'-biphenyl]-4-ol, 4-(4-Chlorophenyl)phenol, 4-(4'-Chlorophenyl)phenol, 4-Chloro-4'-biphenylol
IUPAC Name 4-(4-chlorophenyl)phenol
SMILES Oc1ccc(cc1)c2ccc(Cl)cc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C12H9ClO/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,14H
Formula Weight 204.0342
Chemical Name or Material 4-Chloro-4'-hydroxybiphenyl