Få mere at vide
4-Chloro-4'-fluorobutyrophenone-d4, TRC
Artikelnummer. 30367883
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30367883 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367883 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36634750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloro-4'-fluorobutyrophenone-d4
CAS 1814904-27-2
Molekylær formel C10H6D4ClFO
Mængde 50 mg
Synonym 4-Chloro-1-(4-fluorophenyl-2,3,5,6-d4)-1-butanone, 1-Chloro-4-(4-fluorophenyl-2,3,5,6-d4)-4-oxobutane, 3-(4-Fluorobenzoyl-2,3,5,6-d4)propyl Chloride, 3-(4-Fluorophenylcarbonyl)-1-chloropropane-d4, 3-(p-Fluorobenzoyl)propyl-d4 Chloride, 3-Chloropropyl 4-Fluorophenyl-d4 Ketone, 4-(4-Fluorophenyl)-4-oxobutyl-d4 Chloride, 4-(p-Fluorophenyl)-4-oxobutyl-d4 Chloride, 4-Chloro-1-(p-fluorophenyl)-1-butanone-d4, 4-Chloro-1-(p-fluorophenyl)-1-oxobutane-d4, 4-Chloro-4'-chlorobutyrophenone-d4, 4-Chloro-4'-fluorobutyrophenone-d4, 4-Chloro-p-fluorobutyrophenone-d4, 4-p-Fluorophenyl-4-oxo-1-chlorobutane-d4, 4'-Fluoro-4-chlorobutyrophenone-d4NSC 87082-d4, p-Fluoro-4-chlorobutyrophenone-d4, p-Fluoro-gamma-chlorobutyrophenone-d4, p-Fluoro-ω-chlorobutyrophenone-d4, gamma-Chloro-4-fluorobutyrophenone-d4, gamma-Chloro-p-fluorobutyrophenone-d4, ω-Chloro-4-fluorobutyrophenone-d4, ω-Chloro-p-fluorobutyrophenone-d4, ω-Chloropropyl 4-Fluorophenyl Ketone-d4
InChI formel InChI=1S/C10H10ClFO/c11-7-1-2-10(13)8-3-5-9(12)6-4-8/h3-6H,1-2,7H2/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(C(=O)CCCCl)c([2H])c([2H])c1F
IUPAC navn 4-chloro-1-(2,3,5,6-tetradeuterio-4-fluorophenyl)butan-1-one
Molekylvægt (g/mol) 204.66
Formel vægt 204.0655