Learn More
4-Chloro-3-nitrocinnamic Acid Methyl Ester, TRC
Product Code. 30366653
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30366653 5 g 5g
1 options
Product Code. 30366653 Supplier Toronto Research Chemicals Supplier No. TRCC5971635G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Chloro-3-nitrocinnamic Acid Methyl Ester
CAS 877065-30-0
Molecular Formula C10H8ClNO4
Quantity 5 g
InChI Formula InChI=1S/C10H8ClNO4/c1-16-10(13)5-3-7-2-4-8(11)9(6-7)12(14)15/h2-6H,1H3/b5-3+
SMILES COC(=O)\C=C\c1ccc(Cl)c(c1)[N+](=O)[O-]
IUPAC Name methyl (E)-3-(4-chloro-3-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol) 241.63
Formula Weight 241.0142