missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7025904
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
97-08-5
C6H3Cl2NO4S
256.053
MFCD00007440
SEWNAJIUKSTYOP-UHFFFAOYSA-N
4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa
7323
4-chloro-3-nitrobenzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl
This item is not returnable. View return policy
97-08-5
C6H3Cl2NO4S
256.053
MFCD00007440
SEWNAJIUKSTYOP-UHFFFAOYSA-N
4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa
7323
4-chloro-3-nitrobenzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 61°C
Quantity 5 g
UN Number UN3261
Beilstein 1978600
Sensitivity Moisture sensitive
Formula Weight 256.07
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'