missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-3'-nitroacetophenone, 97%, Thermo Scientific™

Catalog Number p-7022376
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5465-65-6
C8H6ClNO3
199.59
MFCD00007083
YEVPHFIFGUWSMG-UHFFFAOYSA-N
4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
79596
1-(4-chloro-3-nitrophenyl)ethanone
CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
This item is not returnable. View return policy
5465-65-6
C8H6ClNO3
199.59
MFCD00007083
YEVPHFIFGUWSMG-UHFFFAOYSA-N
4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
79596
1-(4-chloro-3-nitrophenyl)ethanone
CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 96°C to 100°C
Quantity 5 g
Beilstein 1640627
Formula Weight 199.6
Percent Purity 97%
Chemical Name or Material 4'-Chloro-3'-nitroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'