Få mere at vide
4-Chloro-3-methylphenol-2,6-d2, TRC
Artikelnummer. 30366975
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30366975 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366975 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3697665MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chloro-3-methylphenol 2,6-d2
CAS 93951-72-5
Molekylær formel C7 2H2 H5 Cl O
Mængde 5 mg
Synonym 4-Chloro-3-methylphenol D2 (2,6-D2), Chlorocresol D2 (phenyl D2), Phen-2,6-d2-ol, 4-chloro-3-methyl- (9CI), 4-Chloro-3-methylphenol (ring-2,6-D2), 4-Chloro-3-methylphenol-2,6-D2, 4-Chloro-2,6-dideuterio-3-methylphenol, Phen-2,6-d2-ol, 4-chloro-3-methyl- (9CI), 4-Chloro-3-methylphenol (ring-2,6-D2), 4-Chloro-3-methylphenol-2,6-D2, 4-Chloro-2,6-dideuterio-3-methylphenol
InChI formel InChI=1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3/i2D,4D
SMIL [2H]c1cc(Cl)c(C)c([2H])c1O
IUPAC navn 4-chloro-2,6-dideuterio-3-methylphenol
Molekylvægt (g/mol) 144.6
Formel vægt 144.0311