missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-hydroxybenzoic Acid Ethyl Ester, TRC

Product Code. 30360675
Change view
Click to view available options
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30360675 100 mg
30352112 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30360675 Supplier Toronto Research Chemicals Supplier No. TRCC366810100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Chloro-3-hydroxybenzoic Acid Ethyl Ester
CAS 335013-90-6
Molekylær formel C9H9ClO3
Mængde 100 mg
Synonym 4-Chloro-3-hydroxybenzoic Acid Ethyl Ester
InChI formel InChI=1S/C9H9ClO3/c1-2-13-9(12)6-3-4-7(10)8(11)5-6/h3-5,11H,2H2,1H3
SMIL CCOC(=O)c1ccc(Cl)c(O)c1
IUPAC navn ethyl 4-chloro-3-hydroxybenzoate
Molekylvægt (g/mol) 200.62
Formel vægt 200.024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.