missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-3-fluorobenzotrifluoride, 98%

Brand:  Alfa Aesar™ H64411.06

Additional Details : CAS Number : 32137-20-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401619

  • 1049.35 DKK / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Chloro-3-fluorobenzotrifluoride
198.54
2093904
32137-20-5
MFCD00060646
1-Chloro-2-fluoro-4-(trifluoromethyl)benzene

Specifications

4-Chloro-3-fluorobenzotrifluoride
135°C
5g
1.4335
98%
MFCD00060646
1-Chloro-2-fluoro-4-(trifluoromethyl)benzene
1.49g/mL
Pale yellow
32137-20-5
Liquid
198.54
2093904
Videos
SDS