missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzoic Acid, TRC

Product Code. 30364896
Change view
Click to view available options
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30364896 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364896 Supplier Toronto Research Chemicals Supplier No. TRCC42770025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Chloro-3-fluorobenzoic Acid
CAS 403-17-8
Molekylær formel C7H4ClFO2
Mængde 25 g
Synonym 4-Chloro-3-fluoro-benzoic Acid, 3-Fluoro-4-chlorobenzoic Acid
InChI formel InChI=1S/C7H4ClFO2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3H,(H,10,11)
SMIL OC(=O)c1ccc(Cl)c(F)c1
IUPAC navn 4-chloro-3-fluorobenzoic acid
Molekylvægt (g/mol) 174.56
Formel vægt 173.9884
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.