missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(ethylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7035458
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871332-69-3
C9H11BClNO3
227.451
MFCD07783861
YMJAUMCTGHBWQK-UHFFFAOYSA-N
4-chloro-3-ethylcarbamoyl phenyl boronic acid, 4-chloro-3-ethylcarbamoyl phenylboronic acid, acmc-209qgz, 4-chloro-3-ethylcarbamoyl phenylboronicacid, 4-chloro-3-ethyl carbamoyl phenylboronic acid, boronic acid,b-4-chloro-3-ethylamino carbonyl phenyl
44886932
[4-chloro-3-(ethylcarbamoyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)Cl)C(=O)NCC)(O)O
This item is not returnable. View return policy
871332-69-3
C9H11BClNO3
227.451
MFCD07783861
YMJAUMCTGHBWQK-UHFFFAOYSA-N
4-chloro-3-ethylcarbamoyl phenyl boronic acid, 4-chloro-3-ethylcarbamoyl phenylboronic acid, acmc-209qgz, 4-chloro-3-ethylcarbamoyl phenylboronicacid, 4-chloro-3-ethyl carbamoyl phenylboronic acid, boronic acid,b-4-chloro-3-ethylamino carbonyl phenyl
44886932
[4-chloro-3-(ethylcarbamoyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)Cl)C(=O)NCC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 220°C to 224°C
Quantity 250 mg
Formula Weight 227.45
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-(ethylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'