missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7056084
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
131748-14-6
C6H3ClF3N
181.54
MFCD07368047
FZINIBTZNSPWQR-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl pyridine, 2-trifluoromethyl-4-chloropyridine, pyridine,4-chloro-2-trifluoromethyl, pubchem15520, acmc-209bnu, 4-chloro-2-trifluoromethyl-pyridine, 2-trifluoro methyl-4-chloro pyridine
14761442
4-chloro-2-(trifluoromethyl)pyridine
FC(F)(F)C1=NC=CC(Cl)=C1
This item is not returnable. View return policy
131748-14-6
C6H3ClF3N
181.54
MFCD07368047
FZINIBTZNSPWQR-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl pyridine, 2-trifluoromethyl-4-chloropyridine, pyridine,4-chloro-2-trifluoromethyl, pubchem15520, acmc-209bnu, 4-chloro-2-trifluoromethyl-pyridine, 2-trifluoro methyl-4-chloro pyridine
14761442
4-chloro-2-(trifluoromethyl)pyridine
FC(F)(F)C1=NC=CC(Cl)=C1

Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 250 mg
UN Number UN2810
Solubility Information Slightly soluble in water.
Formula Weight 181.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'