missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-7055495
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3678-63-5
C6H6ClN
127.571
MFCD00038833
DAOZBJCTEPJGES-UHFFFAOYSA-N
4-chloro-2-picoline, 2-methyl-4-chloropyridine, 4-chloro-2-methyl-pyridine, pyridine, 4-chloro-2-methyl, p-chloropicoline, 4-chloropicoline, 4-chloro-2-picolin, acmc-20amln, pubchem2071
581392
4-chloro-2-methylpyridine
CC1=NC=CC(=C1)Cl
This item is not returnable. View return policy
3678-63-5
C6H6ClN
127.571
MFCD00038833
DAOZBJCTEPJGES-UHFFFAOYSA-N
4-chloro-2-picoline, 2-methyl-4-chloropyridine, 4-chloro-2-methyl-pyridine, pyridine, 4-chloro-2-methyl, p-chloropicoline, 4-chloropicoline, 4-chloro-2-picolin, acmc-20amln, pubchem2071
581392
4-chloro-2-methylpyridine
CC1=NC=CC(=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.15 g/mL
Boiling Point 54°C to 56°C (15 mmHg)(literature)
Refractive Index 1.5205
Quantity 1 g
Beilstein 107196
Formula Weight 127.57
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'