missing translation for 'onlineSavingsMsg'
Få mere at vide

(4-Chloro-2-methylphenoxy-2,3,5-d3)acetic Acid, TRC

Artikelnummer. 30365053
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365053 1 mg
30364194 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365053 Leverandør Toronto Research Chemicals Leverandørnr. TRCC6020051MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 352431-14-2
Molekylær formel C9 2H3 H6 Cl O3
Molekylvægt (g/mol) 203.64
Synonym Acetic acid, (4-chloro-2-methylphenoxy-2,3,5-d3)- (9CI), MCPA-d3, MCPA D3 (phenyl D3)
IUPAC navn 2-(4-chloro-2,3,5-trideuterio-6-methylphenoxy)acetic acid
SMIL [2H]c1c([2H])c(OCC(=O)O)c(C)c([2H])c1Cl

Tekniske data

Kemisk navn eller materiale MCPA D3
Mængde 1 mg
InChI formel InChI=1S/C9H9ClO3/c1-6-4-7(10)2-3-8(6)13-5-9(11)12/h2-4H,5H2,1H3,(H,11,12)/i2D,3D,4D
Formel vægt 203.0429
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.