missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number 11401870
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
55685-75-1
C8H9ClO2
172.61
MFCD02683547
AXXPZAITCCIOIA-UHFFFAOYSA-N
4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
3549393
(4-chloro-2-methoxyphenyl)methanol
COC1=C(CO)C=CC(Cl)=C1
This item is not returnable. View return policy
55685-75-1
C8H9ClO2
172.61
MFCD02683547
AXXPZAITCCIOIA-UHFFFAOYSA-N
4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
3549393
(4-chloro-2-methoxyphenyl)methanol
COC1=C(CO)C=CC(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 46°C to 50°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 172.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methoxybenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'