missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number p-3771079
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
90296-27-8
C8H9ClO2
172.61
MFCD02683547
AXXPZAITCCIOIA-UHFFFAOYSA-N
4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
3549393
(4-chloro-2-methoxyphenyl)methanol
COC1=C(CO)C=CC(Cl)=C1
This item is not returnable. View return policy
90296-27-8
C8H9ClO2
172.61
MFCD02683547
AXXPZAITCCIOIA-UHFFFAOYSA-N
4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
3549393
(4-chloro-2-methoxyphenyl)methanol
COC1=C(CO)C=CC(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 47°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H3(OH)OCH3
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 172.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methoxybenzyl alcohol
compliance-icons
Product Identifier
  • 4-Chloro-2-methoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'